Läs mer
Cortisone Sodium Succinate, TRC
Produktkod. 30360813
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360813 25 mg
30350376 10 mg
30359453 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30360813 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC69650525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7415-42-1
Molekylformel C25H31NaO8
Molekylvikt (g/mol) 482.5
Synonym 21-(Hydrogen Succinate) Cortisone Sodium Salt, 21-(3-Carboxy-1-oxopropoxy)-17-hydroxy-pregn-4-ene-3,11,20-trione Monosodium Salt
IUPAC-namn sodium;4-[2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoate
LEDER [Na+].C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(=O)[O-])[C@@]4(C)CC(=O)[C@H]23
Specifikationer
Kemiskt namn eller material Cortisone Sodium Succinate
Kvantitet 25 mg
InChI formel InChI=1S/C25H32O8.Na/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;/h11,16-17,22,32H,3-10,12-13H2,1-2H3,(H,29,30);/q;+1/p-1/t16-,17-,22+,23-,24-,25-;/m0./s1
Formel vikt 482.1917