Läs mer
Corynoline, TRC
Produktkod. 30366841
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366841 10 mg
30355078 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366841 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC69690010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18797-79-0
Molekylformel C21 H21 N O5
Molekylvikt (g/mol) 367.4
Synonym [1,3]Benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridin-6-ol, 5b,6,7,12b,13,14-hexahydro-5b,13-dimethyl-, (5bR,6S,12bR)-, Chelidonine, 13-methyl-, Corynoline (7CI,8CI), [1,3]Benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridine, chelidonine deriv., (+)-Corynoline, (5bR,6S,12bR)-5b,6,7,12b,13,14-Hexahydro-5b,13-dimethyl[1,3]benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridin-6-ol, [1,3]Benzodioxolo[5,6-c]-1,3-dioxolo[4,5-i]phenanthridin-6-ol, 5b,6,7,12b,13,14-hexahydro-5b,13-dimethyl-, [5bR-(5bα,6β,12bα)]-
IUPAC-namn (1R,12S,13R)-13,24-dimethyl-5,7,18,20-tetraoxa-24-azahexacyclo[11.11.0.02,10.04,8.014,22.017,21]tetracosa-2,4(8),9,14(22),15,17(21)-hexaen-12-ol
LEDER CN1Cc2c3OCOc3ccc2[C@@]4(C)[C@@H](O)Cc5cc6OCOc6cc5[C@@H]14
Specifikationer
Kemiskt namn eller material Corynoline
Kvantitet 10 mg
InChI formel InChI=1S/C21H21NO5/c1-21-14-3-4-15-19(27-10-24-15)13(14)8-22(2)20(21)12-7-17-16(25-9-26-17)5-11(12)6-18(21)23/h3-5,7,18,20,23H,6,8-10H2,1-2H3/t18-,20+,21-/m0/s1
Formel vikt 367.142