Läs mer
Corynoxeine, MedChemExpress
Produktkod. 16449028
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16449028 10 mg
16431448 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16449028 Leverantör Medchem Express Leverantörsnummer HYN0590
MedChemExpress Corynoxeine, isolated from the hook of Uncaria rhynchophylla, is a potent ERK1/ERK2 inhibitor of key PDGF-BB-induced vascular smooth muscle cells (VSMCs) proliferation.
Corynoxeine, isolated from the hook of Uncaria rhynchophylla, is a potent ERK1/ERK2 inhibitor of key PDGF-BB-induced vascular smooth muscle cells (VSMCs) proliferation.
Specifikationer
Kemiskt namn eller material Corynoxeine
CAS 630-94-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C22H26N2O4
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 3.8 mg/mL (9.94 mM)
LEDER O=C(NC1=C2C=CC=C1)[C@]32[C@@](C[C@H](/C(C(OC)=O)=C\OC)[C@@H](C=C)C4)([H])N4CC3
Molekylvikt (g/mol) 382.45
Formel vikt 382.45
Procent renhet 99.58%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation