Läs mer
Corning™ Costar™ Cell Culture Triangular Flasks
Handla allt Corning Produkter
Stängningsfärg:
Black
Orange
Hals stil:
Angled
Straight
Förpackningsstorlek:
Paket med 100
Paket med 200
Paket med 500
Beskrivning
- Medical-grade polystyrene is sterile and nonpyrogenic
- Tissue culture-treated to promote optimal cell attachment and growth
- Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
- Graduated
- Frosted labeling surfaces
- Extra-wide neck openings for easier pipetting
- Printed lot number and catalog number on each flask
- Vented filter caps
- Maximize gas exchange while minimizing the possibility of contamination
- Hydrophobic 0.2 μm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened
Specifikationer
Specifikationer
| Stängningsfärg | Black |
| Stängningsmaterial | HDPE |
| Material | Polystyrene |
| För användning med (applikation) | For optimal cell attachment and growth |
| Hals stil | Angled |
| Form | Triangular |
| Ytbeläggning | Tissue Culture Treated |
| Graderad | Yes |
| Sterilitet | Sterile |