Läs mer
Corning™ Costar™ Cell Culture Triangular Flasks
GREENER_CHOICE
Produktkod. 10762122 Handla allt Corning Produkter
Stängningsfärg:
Black
Orange
Hals stil:
Angled
Straight
Förpackningsstorlek:
Paket med 100
Paket med 200
Paket med 500
Produktkod. 10762122
Brand: Corning™ 3276
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.
  • Medical-grade polystyrene is sterile and nonpyrogenic
  • Tissue culture-treated to promote optimal cell attachment and growth
  • Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
  • Graduated
  • Frosted labeling surfaces
  • Extra-wide neck openings for easier pipetting
  • Printed lot number and catalog number on each flask
  • Vented filter caps
  • Maximize gas exchange while minimizing the possibility of contamination
  • Hydrophobic 0.2 μm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened
TRUSTED_SUSTAINABILITY
Specifikationer
Stängningsfärg Orange
Stängningsmaterial HDPE
Material Polystyrene
För användning med (applikation) For optimal cell attachment and growth
Hals stil Straight
Form Triangular
Ytbeläggning Tissue Culture Treated
Graderad Yes
Sterilitet Sterile