Beskrivning
- For use as reservoir tray with HTS Transwell Permeable Supports
- TC-treated for cell attachment
- Same footprint as standard 24 well plate but with single rectangular well
Specifikationer
Specifikationer
| Stängningsfärg | Clear |
| Förslutning ingår | Yes |
| Stängningsmaterial | Polystyrene |
| Bottenform | Flat |
| Färg | Clear |
| Material | Polystyrene |
| Form | Rectangular |
| Sterilitet | Sterile |
| Ytbehandling | Cell Culture |
| Typ | Cell Culture Plate |
| Visa mer |