Läs mer
Cotrimoxazole, TRC
Produktkod. 30010285
Change view
Kvantitet:
1 g
2.5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010285 25 g
30006219 1 g
30005005 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC72530025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 8064-90-2
Molekylformel C24H29N7O6S
Molekylvikt (g/mol) 543.6
Synonym Abacin, Abactrim, Acuco, Alcorim F, Alfatrim, Andoprim, Anitrim, Antibiotic TMP-SMZ, Antrimox, Aop-Sulfatrim, Apo-Sulfatrim, BS, Bacdan, Bacidal, Bacin, Bacticel, Bactifor, Bactoprim, Bactramin, Bactrim, Bactrim DS, Bactrim Forte, Bactrimel, Baktar, Bencole, Bethaprim, Biosulten, Biseptol, Biseptol 480, Brifeseptol, Briscotrim, Bronquimucil, Centrim, Chemitrim, Chemotrim, Ciplin, Co-Trimoxazole, Co-Trimoxizole, Co-trimox, Colizole, Colizole DS, Comox, Conprim, Cotrim, Cotrim-Diolan, Cotrimel, Cotrimoxazol, Cotrumoxazole, Deprim, Diseptyl, Drylin, Duon, Duratrimet, Ectaprim, Entrim, Esbesul, Espectrin, Euctrim, Eusaprim, Exbesul, Fectrim, Fermagex, Fortrim, Futin, Gantaprim, Gantrim, Groseptol 480, Hulin, Ikaprim, Imexim, Intertrim, Isotrim, Kemoprim Forte, Kepinol, Kepinol forte, Lagatrim, Lagatrim Forte, Laratrim, Lastrim, Lescot, Linaris, Medixin, Microtrim, Nopil, Novotrimel, Omsat, Omstat, Oraprim, Oriprim, Ortizol Oblet, Oxaprim, Pantoprim, Purbal, Resprim, Resprim Forte, Roubal, SMX-TMP, SMZ-TMP, SXT, Salvatrim, Septazole, Septra, Septran, Septrim, Septrin, Septrin DS, Septrin Forte, Septrin S, Setprin, Sigaprim, Stopan, Sugaprim, Sulfacet, Sulfamar, Sulfamethoprim, Sulfamethoxazole-trimethoprim mixt., Sulfaprim, Sulfatrim, Sulfatrimethoprim, Sulfazotrim, Sulfazotrin, Sulfinam, Sulfotrim, Sulfotrimin, Sulfoxaprim, Sulmeprim, Sulprim, Sulthrim, Sultrex, Sumetrolim, Supracombin, Suprim, Suprin, Sutrim, TMP-SMX, TMP-SMZ, TMS, Teleprim, Thiocuran, Trasepton, Trigonyl, Trim, Trim-Sulfa, Trimel, Trimesulf, Trimeth/sulfa, Trimethazol, Trimethoprim-sulfamethoxazole mixt., Trimethoprimsulfa, Trimetox, Trimezol, Trimezole, Trimosulfa, Trisul, Trisulcom, Trisulfam, Trisural, Trizole, Ulfaprim, Uro-D S, Urobactrim, Uroplus, Xeroprim
IUPAC-namn 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide;5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
LEDER CCOC(=O)C(C)NCCCC[C@H](N)C(=O)OC
Specifikationer
Kemiskt namn eller material Cotrimoxazole
Anteckningar om renhetsgrad HPLC
Kvantitet 25 g
InChI formel InChI=1S/C14H18N4O3.C10H11N3O3S/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15;1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h5-7H,4H2,1-3H3,(H4,15,16,17,18);2-6H,11H2,1H3,(H,12,13)
Procent renhet >95%