Läs mer
Coumaphos, TRC
Produktkod. 30361339
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361339 500 mg
30360856 1 g
30356024 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30361339 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC745300500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56-72-4
Molekylformel C14 H16 Cl O5 P S
Molekylvikt (g/mol) 362.77
Synonym Coumarin, 3-chloro-7-hydroxy-4-methyl-, O-ester with O,O-diethyl phosphorothioate (8CI), Umbelliferone, 3-chloro-4-methyl-, O,O-diethyl phosphorothioate (6CI), 3-Chloro-4-methyl-7-coumarinyl diethyl phosphorothioate, 3-Chloro-4-methyl-7-hydroxycoumarin diethyl thiophosphoric acid ester, 3-Chloro-4-methylumbelliferone O-ester with O,O-diethyl phosphorothioate, 3-Chloro-7-hydroxy-4-methyl-coumarin O,O-diethyl phosphorothioate, 3-Chloro-7-hydroxy-4-methyl-coumarin O-ester with O,O-diethyl phosphorothioate, Agridip, Asuntol, Bayer 21/199, Baymix, Baymix 50, Co-ral, Co-ral (phosphate), Coumafos, Coumaphos, ENT 17,957, Koral, Meldane, Meldone, Muscatox, Muscotox, NSC 8944, O,O-Diethyl 3-chloro-4-methyl-7-umbelliferone thiophosphate, O,O-Diethyl O-(3-chloro-4-methyl-2-oxo-2H-benzopyran-7-yl) phosphorothioate, O,O-Diethyl O-(3-chloro-4-methyl-7-coumarinyl) phosphorothioate, O,O-Diethyl O-3-chloro-4-methyl-2-oxo-2H-1-benzopyran-7-yl phosphorothioate, O-(3-Chloro-4-methyl-2-oxo-2H-1-benzopyran-7-yl) O,O-diethyl phosphorothioate, O-3-Chloro-4-methyl-7-coumarinyl O,O-diethyl phosphorothioate, Perizin, Prince, Resitox
IUPAC-namn 3-chloro-7-diethoxyphosphinothioyloxy-4-methylchromen-2-one
LEDER CCOP(=S)(OCC)Oc1ccc2C(=C(Cl)C(=O)Oc2c1)C
Specifikationer
Kemiskt namn eller material Coumaphos
Kvantitet 500 mg
InChI formel InChI=1S/C14H16ClO5PS/c1-4-17-21(22,18-5-2)20-10-6-7-11-9(3)13(15)14(16)19-12(11)8-10/h6-8H,4-5H2,1-3H3
Formel vikt 362.0145