Learn More
Cole-Parmer™ Coupling miniature quick disconnect Male Coupler w/Hose Barb
Miniature quick-disconnect couplings are able to offer a more secure connection than Luer fittings.
Brand:  Cole-Parmer™ 06363-58
Additional Details : Weight : 0.01000kg
Product Code. 15456473
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Miniature quick-disconnect couplings are able to handle 1/16 in. ID tubing and offer a more secure connection than luer fittings.
  • Fitting halves engage and seal with a twist.
  • Once engaged, the coupling allows the tubing to rotate freely to help prevent kinked tubing and accidental disconnections.
Specifications
Specifications
Hose barb Coupler, Male, 1/16 in., 10/pk (Polypropylene)
Polypropylene
3/32 in.
Connection type: Quick-Disconnect x Barb
1/16 in.
10/pk.
Ethylene Oxide(ETO) or gamma