Learn More
Cole-Parmer™ Coupling miniature quick disconnect Male Coupler w/Hose Barb
Miniature quick-disconnect couplings are able to offer a more secure connection than Luer fittings.
Brand: Cole-Parmer™ 06363-58
Additional Details : Weight : 0.01000kg
Description
- Miniature quick-disconnect couplings are able to handle 1/16 in. ID tubing and offer a more secure connection than luer fittings.
- Fitting halves engage and seal with a twist.
- Once engaged, the coupling allows the tubing to rotate freely to help prevent kinked tubing and accidental disconnections.
Specifications
Hose barb Coupler, Male, 1/16 in., 10/pk (Polypropylene) | |
Polypropylene | |
3/32 in. | |
Connection type: Quick-Disconnect x Barb |
1/16 in. | |
10/pk. | |
Ethylene Oxide(ETO) or gamma |