missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Coupling standard quick disconnect Female Coupler w/Hose Barb

Standard flow quick-disconnect female coupler with hose barb.

Brand:  Masterflex™ 06364-65

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15436563

  • 166.00 SEK / Each


Estimated Shipment
30-12-2020
Add to basket

Description

Description

  • Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
  • Plug insert into the body to connect fitting—fitting halves lock into place immediately.
  • To disconnect fitting halves, simply press the thumb latch on the coupling body.
Specifications

Specifications

120 psi
Yes
5.0038 cm
Autoclave or ethylene oxide (ETO)
Quick-Disconnect Hose Barb Fitting Body; Valved, 1/4 in. Tube ID, 1/4 in. Flow Size
Polypropylene, Valved
1/4 in. Tubing
EPR
Videos
SDS
Documents