Läs mer
Epredia™ Täckglas, rektanglar
Produktkod. p-7112203 Handla allt Epredia Produkter
Tjocklek (metrisk):
#1 (0,13 till 0,16 mm)
#1,5 (0,16 till 0,19 mm)
Mått (L x B):
36 x 24 mm
64 x 48 mm
Förpackningsstorlek:
Paket med 1000
Varje
Produktkod. 12765098
Brand: Epredia™ A67761330
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Epredia™TM coverslips are ideally suited as cover material for specimens in microscopic examinations in medicine, biology and research. Manufactured from D 263TM M borosilicate glass from Schott AG. Available in various sizes and thicknesses.
  • Highly transparent and achromatic – practically free of any color distortion
  • Good planarity
  • Refraction index adjusted for microscopes
  • High optical homogeneity – practically free of inclusions, streaks, bubbles etc.
  • Very good chemical stability
TRUSTED_SUSTAINABILITY
Specifikationer
Form Rectangle
Längd (metrisk) 64 mm
Bredd (metrisk) 48 mm
Tjocklek (metrisk) #1 (0,13 till 0,16 mm)
Mått (L x B) 64 x 48 mm
Kvantitet 100/Pk.