missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11464-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Cytochrome c oxidase 17 (COX17) is a nuclear gene encoding an assembly protein necessary for proper COX apoenzyme-dependent mitochondrial respiration. The mammalian COX apoenzyme is a heteromer consisting of three mitochondrial genes that encode catalytic subunits and several nuclear genes that encode structural subunits. COX17 influences the recruitment of copper to the mitochondria for incorporation into the COX apoenzyme.Specifications
COX17 | |
Polyclonal | |
Unconjugated | |
COX17 | |
COX17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10063, 12856, 89786 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P56394, Q14061 | |
Cox17 | |
COX17 Fusion Protein Ag2032 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |