missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ COX7A2L Recombinant Protein

Brand:  Abnova™ H00009167-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16163352

  • 6845.00 SEK / 25µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH05251.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
38.17
8
Glutathione Sepharose 4 Fast Flow
25 ug
MYYKFSGFTQKLAGAWASEAYSPQGLKPVVSTEAPPIIFATPTKLTSDSTVYDYAGKNKVPELQKFFQKADGVPVYLKRGLPDQMLYRTTMALTVGGTIYCLIALYMASQPKNK
COX7AR/COX7RP/EB1/SIG81
COX7A2L
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
9167
COX7A2L (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COX7A2L
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers