missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ CP 809101 hydrochloride

Description
Potent and selective 5-HT2C receptor agonist (pEC50 values are 9.96, 7.19 and 6.81 for human 5-HT2C, 5-HT2B and 5-HT2A receptors respectively). Displays antipsychotic activity; suppresses condition avoidance responding (CAR) and inhibits PCP and amphetamine-stimulated hyperactivity in rats following subcutaneous administration.
Specifications
Specifications
| Kemiskt namn eller material | CP 809101 hydrochloride |
| Kvantitet | 10 mg |
| Formel vikt | 341.24 |
| Renhet | >99% |
| Molekylvikt (g/mol) | 341.236 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?