Learn More
Cole-Parmer™ CPC™ High-Flow Quick-Disconnect Fitting with Bulkhead Panel Mount Hose Barb Bodies
Eliminate leaks with these valved couplings
Brand:  Cole-Parmer™ 31307-25
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15497123
  • 242.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately).
  • Plug insert into coupling body to connect.
  • Fitting halves lock into place immediately.
  • To disconnect fitting halves, simply press the button on the coupling body.
  • Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged.
  • All fitting bodies have 316 SS latch springs and EPDM seals.
Specifications
Specifications
Grey
Yes
Valved Bulkhead Panel Mount Hose Barb Body
137°C
-40°C
27.5 in. Hg
3/8 in. Quick Disconnect
Quick-Disconnect x Barb Connection
Polysulfone
125psi
-40°F
3/8 in.
3/8 in.
Autoclave, Ethylene Oxide (ETO) or Gamma
Videos