Learn More
Cole-Parmer™ CPC™ Quick Disconnect Non-valved Polypropylene Pipe Adapter Bodies
Quick Disconnect couplings feature a thumb latch for easy assembly
Brand:  Cole-Parmer™ 31303-68
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15497083
  • 194.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately)
  • Plug insert into coupling body to connect
  • Fitting halves lock into place immediately
  • To disconnect fitting halves, simply press the button on the coupling body
  • Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged
  • All fitting bodies have 316 SS latch springs and EPDM seals
Specifications
Specifications
No
Non-valved Polypropylene Pipe Adapter Bodies
71°C
0°C
27.5 in. Hg
Gamma or e-beam
Polypropylene
60psi
32°F
1/2 in.
3/8 in. Quick-Disconnect
Videos