Learn More
R&D Systems™ CpG oligodeoxynucleotides Mouse
Product Code. 18174724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
Product Code. Quantity Unit Size
18174724 0.025 mg 25µg
1 options
Product Code. 18174724 Supplier R&D Systems™ Supplier No. NBP231141SS
  • ONLINE EXCLUSIVE PRICE
    2075.83 SEK
    1747.20 SEK / 25µg
    Save 328.63 SEK
    15% Av
 Restricted Item
Beräknad leverans: 04-12-2026
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 0.025 mg
For Use With (Application) Activation of mouse TLR9
Type CpG Oligodeoxynucleotides