Läs mer
CPI 613, TRC
Produktkod. 17992935
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17992935 5 mg
17952075 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17992935 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC7814405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 95809-78-2
Molekylformel C22H28O2S2
Molekylvikt (g/mol) 388.59
Synonym 6,8-Bis[(phenylmethyl)thio]-octanoic Acid, 6,8-Bis(benzylthio)-octanoic Acid
IUPAC-namn 6,8-bis(benzylsulfanyl)octanoic acid
LEDER OC(=O)CCCCC(CCSCc1ccccc1)SCc2ccccc2
Specifikationer
Kemiskt namn eller material CPI 613
Kvantitet 5 mg
InChI formel InChI=1S/C22H28O2S2/c23-22(24)14-8-7-13-21(26-18-20-11-5-2-6-12-20)15-16-25-17-19-9-3-1-4-10-19/h1-6,9-12,21H,7-8,13-18H2,(H,23,24)