missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPLX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10246-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CPLX1 encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release.Specifications
CPLX1 | |
Polyclonal | |
Unconjugated | |
CPLX1 | |
complexin 1, Complexin I, complexin1, CPLX1, CPX I, CPX1, Synaphin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10815, 12889, 64832 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14810, P63040, P63041 | |
Cplx1 | |
CPLX1 Fusion Protein Ag0387 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |