missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Mouse anti-Human, Unconjugated, Clone: 2A12A4, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CPS1 |
---|---|
Clone | 2A12A4 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840366
|
Proteintech
67630-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830366
|
Proteintech
67630-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CPS1 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human CPS1. This antibody recognizes human, mouse, rat antigen. The CPS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CPS1 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1373 | |
Primary |
2A12A4 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |