missing translation for 'onlineSavingsMsg'
Learn More

CPSF4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15023-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883994

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

LUZP1 encodes a protein that contains a leucine zipper motif. The exact function of the encoded protein is not known. In mice this gene affects neural tube closure.
Specifications

Specifications

CPSF4
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95639, Q8BQZ5
CPSF 30 kDa subunit, CPSF30, CPSF4, NAR, Neb 1, NEB1, No arches homolog
Rabbit
IgG
20 μL
-20°C
Primary
10898, 54188
Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
CPSF4
Liquid
Cpsf4
CPSF4 Fusion Protein Ag7103
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers