Learn More
CPT1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22170-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and one of the transcripts is bicistronic.Specifications
CPT1B | |
Polyclonal | |
Unconjugated | |
CPT1B | |
CPT I, CPT1 M, CPT1B, CPTI M, KIAA1670, M CPT1, MCCPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12895, 1375, 25756 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q63704, Q924X2, Q92523 | |
CPT1B | |
CPT1B-specific Fusion Protein Ag17840 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |