missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A5F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66468-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.Specifications
CRABP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P22935, P29373, P51673 | |
CRABP2 | |
CRABP2 Fusion Protein Ag0309 | |
20 μL | |
Primary | |
Human, Porcine, Rat, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A5F3 | |
Unconjugated | |
CRABP2 | |
CRABP II, CRABP2, RBP6 | |
Mouse | |
Protein G | |
RUO | |
100155151, 12904, 1382, 29563 | |
-20°C | |
Liquid |