Learn More
Abnova™ CRADD Recombinant Protein
Human full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00008738-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Encoded by CRADD
- Death domain (CARD/DD) containing protein
- Molecular weight: 47.63kDa
- Preparation method: in vitro wheat germ expression system
- Purification: glutathione Sepharose 4 fast flow
- Storage buffer: 50 mM Tris-HCI, 10 mM reduced glutathione
- pH=8.0 in elution buffer
Sequence: MEARDKQVLRSLRLELGAEVLVEGLVLQYVYEEGILTENHIQEINAQTTGLRKTMLLLDILPSRGPKAFDTFLDSLQEFPWVREKLKKAREEAMTDLPAGDRLTGIPSHILNSSPSDRQINQLAQRLGPEWEPMVLSLGLSQTDIYRCKANHPHNVQSQVVEAFIRWRQRFGKQATFQSLHNGLRAVEV
DPSLLLHMLE
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH37905 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
47.63 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MEARDKQVLRSLRLELGAEVLVEGLVLQYVYEEGILTENHIQEINAQTTGLRKTMLLLDILPSRGPKAFDTFLDSLQEFPWVREKLKKAREEAMTDLPAGDRLTGIPSHILNSSPSDRQINQLAQRLGPEWEPMVLSLGLSQTDIYRCKANHPHNVQSQVVEAFIRWRQRFGKQATFQSLHNGLRAVEVDPSLLLHMLE | |
MGC9163/RAIDD | |
CRADD | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
8738 | |
CRADD (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CRADD | |
Human | |
Recombinant | |
Solution |