missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine phosphate, disodium salt, 99.12%, MP Biomedicals™
Click to view available options
Quantity:
1 g
25 g
Specifications
Color | White |
Physical Form | Crystalline Powder |
Quantity | 1 g |
Assay Percent Range | 98-100% |
Formula Weight | 327.2 |
Chemical Name or Material | Creatine phosphate, disodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'