missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB3L1 OASIS Mouse anti-Human, Unconjugated, Clone: 2B5B4, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CREB3L1 OASIS |
---|---|
Clone | 2B5B4 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840346
|
Proteintech
67617-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830346
|
Proteintech
67617-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CREB3L1, OASIS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CREB3L1, OASIS. This antibody recognizes human, mouse, rat antigen. The CREB3L1, OASIS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CREB3L1 OASIS | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
90993 | |
Primary |
2B5B4 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |