Läs mer
Crenolanib Trifluoroacetic Acid Salt, TRC
Produktkod. 30368559
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30368559 5 mg
1 options
Produktkod. 30368559 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC7818005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Crenolanib Trifluoroacetic Acid Salt
CAS 1797008-57-1
Molekylformel C28H30F3N5O4
Kvantitet 5 mg
Synonym 1-[2-[5-[(3-Methyl-3-oxetanyl)methoxy]-1H-benzimidazol-1-yl]-8-quinolinyl]-4-piperidinamine Trifluoroacetic Acid Salt, ARO 002 Trifluoroacetic Acid Salt, CP 868596 Trifluoroacetic Acid Salt, [1-[2-[5-(3-Methyloxetan-3-ylmethoxy)benzimidazol-1-yl]quinolin-8-yl]piperidin-4-yl]amine Trifluoroacetic Acid Salt
InChI formel InChI=1S/C26H29N5O2.C2HF3O2/c1-26(14-32-15-26)16-33-20-6-7-22-21(13-20)28-17-31(22)24-8-5-18-3-2-4-23(25(18)29-24)30-11-9-19(27)10-12-30;3-2(4,5)1(6)7/h2-8,13,17,19H,9-12,14-16,27H2,1H3;(H,6,7)
LEDER CC1(COc2ccc3c(c2)ncn3c4ccc5cccc(N6CCC(N)CC6)c5n4)COC1.OC(=O)C(F)(F)F
IUPAC-namn 1-[2-[5-[(3-methyloxetan-3-yl)methoxy]benzimidazol-1-yl]quinolin-8-yl]piperidin-4-amine;2,2,2-trifluoroacetic acid
Molekylvikt (g/mol) 557.56