missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol Red Indicator, ≥83%, Honeywell Fluka™
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
UN Number | NONH for all modes of transport |
Beilstein | 343399 |
Formula Weight | 382.43g/mol |
Loss on Drying | 0 to 5% |
Packaging | Glass Bottle |
Density | 0.998g/cm3 |
Color | Brown/Red |
pH | 7.2 to 8.8 (yellow to red) |
Quantity | 10 g |
Physical Form | Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'