Läs mer
Grant Instruments™ CRF-1 - Controlled Rate Freezer
Produktkod. p-7224813
Change view
Modell:
CRF-1 H01
CRF-1 H05
CRF-1 H06
Håller:
Djupa kryoflaskor (55 x 1,8 ml)
IMV-sugrör (18 x 0,3 ml)
SBS-mikroplatta (1)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Modell Håller
16389346 CRF-1 H01 IMV-sugrör (18 x 0,3 ml)
15982741 CRF-1 H05 SBS-mikroplatta (1)
16369646 CRF-1 H06 Djupa kryoflaskor (55 x 1,8 ml)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16389346 Leverantör Grant Instruments™ Leverantörsnummer CRF1 H01
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CRF-1 Liquid nitrogen free and cryogen free controlled rate freezer for research into the cryopreservation of a wide range of material including: embryos, stem cells, mammalian cells, spermatozoa, antibodies, tissue sections and rodent organs.
CRF-1 Liquid nitrogen free and cryogen free controlled rate freezer for research into the cryopreservation of a wide range of material including: embryos, stem cells, mammalian cells, spermatozoa, antibodies, tissue sections and rodent organs. The CRF-1 brings accuracy, precision and reproducibility to biological cryopreservation.
Unlike conventional liquid nitrogen based controlled rate cooling equipment, the CRF-1 poses no contamination risk and can be used in cleanrooms and barrier facilities. The CRF1 fits neatly and quietly on a bench-top and its performance in terms of cell viability after freezing is comparable or better than standard liquid nitrogen freezers. As alcohol is not used, there is also no potential fire risk. The CRF-1 will cool down to -100°C with straws.
The cooling rate of the CRF-1 is precisely controlled, ensuring accuracy and reproducibility throughout the freezing profile, especially for the important nucleation/seeding phase. This ensures optimal recovery of cells upon thawing. Operation is simple and can be carried out with or without a PC; data can be logged via PC software and cooling profiles are directly displayed. Different cooling profiles are available from a drop down menu and customised profiles can be written.
Main applications:
The CRF-1 is highly versatile and can be used for the cryopreservation research of a wide range
of samples in cryovials, straws, bags, microplates and Matrix-96-well block plates in the following
areas:
• Transgenic embryos research
• Stem cell research
• Clinical and research samples, e.g. lymphocytes and tissue cell lines in conventional cryovials
• Various mammalian cells including cardiomyocytes, adipose, liver and muscle
• Cord blood derived stem cells
• Adherent cells and stem cells in microplates
• Cell suspensions in numbered/barcoded arrays
• Robotic integration – the CRF-1 has also successfully been integrated into robotic systems
• Suitable for applications in veterinarian IVF
Key benefits and features:
• Accurate and reproducible control of cooling rates and sample temperatures
• Easy to use and samples can be nucleated/seeded in-situ
• Linear and non-linear cooling profiles
• Low running costs: estimated at 1% of liquid nitrogen-controlled rate freezing
• Temperature remains at -100°C at the end of cycle for straw applications until freezer
is switched off
• Un-interruptible Power Supply (UPS): complete cycle run if power fails (supplied as an
optional accessory)
• CE marked (laboratory use)
• Servicing and calibration available
• 3 year warranty
Dimensions (w x d x h): 264 x 367 x 445 mm
Weight (Kg): 9.8
Max plate temperature: 30°C
Min plate temperature: -80°C or -100°C (dependent on model)
Temperature accuracy: ± 0.5°C
Temperature stability: ± 0.1ºC
Temperature uniformity: ± 1.0°C
Max cooling rate: 10C°C /min
Max temperature stabilization time: 5 minutes
TRUSTED_SUSTAINABILITY
Specifikationer
Typ CRF-1 H01 Controlled rate freezer for 18 x 0.3 mL IMV straws.
Modell CRF-1 H01
Vikt (metrisk) 34 kg
Håller IMV-sugrör (18 x 0,3 ml)