Learn More
DWK Life Sciences Crimp Top Vial
Crimp top vials are the product of choice for many routine high performance liquid chromatography (HPLC) and gas chromatography (GC) applications.
Brand: DWK Life Sciences 225602
Additional Details : Weight : 0.26000kg
Description
Chromatography vials can be distinguished based on size and finish (thread, crimp, snap). They come in clear and amber glass and with or without writing patch. The most common vial sizes for liquid chromatography vials are 12 x 32 mm as well as 15 x 45 mm. Depending on the resource, 12 x 32 mm vials will also be described as 1.5 mL, 1.8 mL or 2.0 mL vials.
- Crimp top vials provide an excellent seal for long term sample storage and analyses involving high volatile solvents
- 40% larger opening improves sample accessibility and reduces the risk of autosampler needle damage
- Packed in re-closable, tamper-proof evident PP-boxes for protection against contamination during transportation and storage
- Optional white ceramic writing patch and fill marks for easy labeling
Specifications
Clear | |
Borosilicate Glass (5.1) | |
High Performance Liquid Chromatography (HPLC), Gas Chromatography (GC) | |
32 mm | |
0.9 mm | |
Yes |
11.6 mm | |
1.5 mL | |
Autoclavable | |
100 | |
ND11 |