Läs mer
CRL-40,941, TRC
Produktkod. 30358987
Change view
Kvantitet:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358987 5 g
30357344 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358987 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC7900005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[[Bis(4-fluorophenyl)methyl]sulfinyl]-N-hydroxy-acetamide
CAS 90212-80-9
Molekylformel C15 H13 F2 N O3 S
Kvantitet 5 g
Synonym CRL-40,941, 2-[[Bis(4-fluorophenyl)methyl]sulfinyl]-N-hydroxy-acetamide
InChI formel InChI=1S/C15H13F2NO3S/c16-12-5-1-10(2-6-12)15(22(21)9-14(19)18-20)11-3-7-13(17)8-4-11/h1-8,15,20H,9H2,(H,18,19)
LEDER ONC(=O)CS(=O)C(c1ccc(F)cc1)c2ccc(F)cc2
IUPAC-namn 2-[bis(4-fluorophenyl)methylsulfinyl]ethanehydroxamic acid
Molekylvikt (g/mol) 325.33
Formel vikt 325.0584