Läs mer
Crocin, TRC
Produktkod. 17978314
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17978314 100 mg
17939554 10 mg
17939924 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17978314 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC794945100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42553-65-1
Molekylformel C44 H64 O24
Molekylvikt (g/mol) 976.96
Synonym β-D-Glucopyranose, 6-O-β-D-glucopyranosyl-, 1,1'-[(2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethyl-2,4,6,8,10,12,14-hexadecaheptaenedioate], 8,8'-Diapo-ψ,ψ-carotenedioic acid, bis(6-O-β-D-glucopyranosyl-β-D-glucopyranosyl) ester, Crocin (7CI), Crocetin bis(gentiobiosyl) ester, Crocetin di(β-D-gentiobiosyl) ester, Crocetin di-β-gentiobiosyl ester, Crocetin digentiobioside, Crocin 1, Crocin 4, Crocin A, Crocin I, all-trans-Crocetin di-β-D-gentiobiosyl ester, trans-Crocetin di(β-D-gentiobiosyl) ester, trans-Crocin-1, α-Crocin
IUPAC-namn bis[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl] (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioate
LEDER C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C(=O)O[C@@H]1O[C@H](CO[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)\C=C\C=C(/C)\C(=O)O[C@@H]3O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O
Specifikationer
Kemiskt namn eller material Crocin
Kvantitet 100 mg
InChI formel InChI=1S/C44H64O24/c1-19(11-7-13-21(3)39(59)67-43-37(57)33(53)29(49)25(65-43)17-61-41-35(55)31(51)27(47)23(15-45)63-41)9-5-6-10-20(2)12-8-14-22(4)40(60)68-44-38(58)34(54)30(50)26(66-44)18-62-42-36(56)32(52)28(48)24(16-46)64-42/h5-14,23-38,41-58H,15-18H2,1-4H3/b6-5+,11-7+,12-8+,19-9+,20-10+,21-13+,22-14+/t23-,24-,25-,26-,27-,28-,29-,30-,31+,32+,33+,34+,35-,36-,37-,38-,41-,42-,43+,44+/m1/s1
Formel vikt 976.3788