Läs mer
Crotethamide, TRC
Produktkod. 30132509
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30132509 2.5 g 2.5g
30008319 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132509 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC8185002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6168-76-9
Molekylformel C12H22N2O2
Molekylvikt (g/mol) 226.32
Synonym 2-Butenamide, N-[1-[(dimethylamino)carbonyl]propyl]-N-ethyl-, Crotonamide, N-[1-(dimethylcarbamoyl)propyl]-N-ethyl- (6CI,7CI,8CI), N-[1-[(Dimethylamino)carbonyl]propyl]-N-ethyl-2-butenamide, Crotetamide, Crotethamid, Crotethamide, Ethylbutamide, α-(N'-Crotonyl-N'-ethylamino)-N,N-dimethylbutyramide
IUPAC-namn 2-[[(E)-but-2-enoyl]-ethylamino]-N,N-dimethylbutanamide
LEDER CCC(N(CC)C(=O)\C=C\C)C(=O)N(C)C
Specifikationer
Kemiskt namn eller material Crotethamide
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C12H22N2O2/c1-6-9-11(15)14(8-3)10(7-2)12(16)13(4)5/h6,9-10H,7-8H2,1-5H3/b9-6+
Formel vikt 226.1681
Procent renhet >95%