missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25476-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles.Specifications
CRTC3 | |
Polyclonal | |
Unconjugated | |
CRTC3 | |
CRTC3, TORC 3, TORC3, Transducer of CREB protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64784, 70461 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6UUV7, Q91X84 | |
CRTC3 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |