missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CRTC3 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824055
|
Proteintech
25476-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814055
|
Proteintech
25476-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles.Specifications
CRTC3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6UUV7, Q91X84 | |
64784, 70461 | |
Peptide | |
Primary | |
-20°C | |
CRTC3 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CRTC3, TORC 3, TORC3, Transducer of CREB protein 3 | |
CRTC3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |