Läs mer
Eppendorf™ CryoCube™ F101h ULT Freezer, with CO2-Backup
Pluggtyp:
Europa
Beskrivning
- Vacuum insulation panels allow for increased internal capacity with the same footprint as traditionally foamed freezers
- Inner doors are gasketed as well as insulated creating 2 separate compartments minimizing cold air loss while you access your samples
- Easy-to-read LED control panel and alarm status
- Special vacuum vent with plunger ergonomically located to gain immediate access to your freezer
- Inner doors can be quickly removed without the need for tools for easier and faster defrosting
- Full stainless steel interior for easy cleaning during maintenance and defrost cycles
- Powder-coated steel exterior
- Special low-temperature outer door seals allow gaskets to remain flexible at –86 °C and prevent ice buildup for easy access to samples
- Password protection of alarm and temperature setpoints to prevent unauthorized setting changes
Specifikationer
Specifikationer
| Kapacitet (metrisk) | 101 L |
| Temperaturområde | -50°C to -86°C |
| Antal dörrar | 1 |
| Innerdörrar | 2 |
| Skåpsmaterial | Stainless Steel |
| Isolering | Vacuum, Polyurethane Foam |
| Interiör | stainless steel |
| Exteriör finish | Powder-coated Steel |
| Antal ställ per sats | 6 |
| Stil | Upright |
| Visa mer |