Läs mer
Sperian™ Honeywell™ Cryogenic Gloves
Beskrivning
- Water-repellant silicone cowhide grain leather
- Gunn cut, winged thumb, crispin style
- Reinforcement between thumb and index finger
- 200mm split leather cuff
- Velcro™ adjustment strap on side of the palm
- Cuff offers good forearm protection against mechanical hazards and limits any risk of intrusion
Specifikationer
Specifikationer
| Produkttyp | Cryogenic Gloves |
| Storlek | 11 |
| Tjocklek (metrisk) | 1.1/1.3 mm |
| Motståndskraftig mot | Resistance to conductive cold: 2, Resistance to contact cold: 2 |
| Tear Rating | 2 |
| Längd (metrisk) | 400 mm |