missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CSF2RA (Human) Recombinant Protein
Human CSF2RA full-length ORF ( AAH02635.1, 1 a.a. - 400 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001438-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLLLVTSLLLCELPHPAFLLIPEKSDLRTVAPASSLNVRFDSRTMNLSWDCQENTTFSKCFLTDKKNRVVEPRLSNNECSCTFREICLHEGVTFEVHVNTSQRGFQQKLLYPNSGREGTAAQNFSCFIYNADLMNCTWARGPTAPRDVQYFLYIRNSKRRREIRCPYYIQDSGTHVGCHLDNLSGLTSRNYFLVNGTSREIGIQFFDSLLDTKKIERFNPPSNVTVRCNTTHCLVRWKQPRTYQKLSYLDFQYQLDVHRKNTQPGTENLLINVSGDLENRYNFPSSEPRAKHSVKIRAADVRILNWSSWSEAIEFGSDDGNLGSVYIYVLLIVGTLVCGIVLGFLFKRFLRIQRLFPPVPQIKDKLNDNHEVEDEIIWEEFTPEEGKGYREEVLTVKEIT
Specifications
AAH02635.1 | |
colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSF2RA | |
GST |
1438 | |
Wheat germ expression system | |
10 μg | |
CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, MGC3848, MGC4838 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |