missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSHL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548503
Additional Details : Weight : 0.01000kg
Description
CSHL1 Polyclonal antibody specifically detects CSHL1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CSHL1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q14406 | |
CSHL1 | |
KLH conjugated synthetic peptide between 156-185 amino acids from the C-terminal region of human CSHL1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CSHL1 | |
chorionic somatomammotropin CS-5; chorionic somatomammotropin hormone like 1; chorionic somatomammotropin hormone-like 1; Chorionic somatomammotropin-like; CS-5; CSHL1; CSHP1; CSL; GHB4; growth hormone B4; growth hormone cluster; hCS-L; Lactogen-like | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1444 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |