missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK1A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55192-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to CSNK1A1.
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling. Phosphorylates CTNNB1 at ′Ser-45′. May phosphorylate PER1 and PER2. May play a role in segregating chromosomes during mitosis.Specifications
CSNK1A1 | |
Polyclonal | |
Unconjugated | |
CSNK1A1 | |
casein kinase 1, alpha 1, Casein kinase I isoform alpha, CK1, CKI alpha, CSNK1A1, HLCDGP1, PRO2975 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113927, 1452, 93687 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48729, P97633, Q8BK63 | |
Csnk1a1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |