missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK1A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CSNK1A1 |
---|---|
Concentration | 0.45 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826685
|
Proteintech
55192-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816685
|
Proteintech
55192-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to CSNK1A1.
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling. Phosphorylates CTNNB1 at ′Ser-45′. May phosphorylate PER1 and PER2. May play a role in segregating chromosomes during mitosis.Specifications
CSNK1A1 | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P48729, P97633, Q8BK63 | |
113927, 1452, 93687 | |
Peptide | |
Primary | |
-20°C | |
CSNK1A1 |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
casein kinase 1, alpha 1, Casein kinase I isoform alpha, CK1, CKI alpha, CSNK1A1, HLCDGP1, PRO2975 | |
Csnk1a1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |