missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
CSNK2A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10992-1-AP-20UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Catalytic subunit of a constitutively active serine/threonine-protein kinase complex that phosphorylates a large number of substrates containing acidic residues C-terminal to the phosphorylated serine or threonine. Regulates numerous cellular processes, such as cell cycle progression, apoptosis and transcription, as well as viral infection. May act as a regulatory node which integrates and coordinates numerous signals leading to an appropriate cellular response. During mitosis, functions as a component of the p53/TP53-dependent spindle assembly checkpoint (SAC) that maintains cyclin-B-CDK1 activity and G2 arrest in response to spindle damage.Specifikationer
| CSNK2A1 | |
| Polyclonal | |
| Unconjugated | |
| CSNK2A1 | |
| Casein kinase II subunit alpha, CK II alpha, CK2a, CK2A1, CK2 alpha, CKII, CSNK2A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116549, 12995, 1457 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P19139, P68400, Q60737 | |
| Csnk2a1 | |
| CSNK2A1 Fusion Protein Ag1452 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering