missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CSNK2A2 (CK2 α 2) Recombinant Human Protein

Casein kinase II is a protein serine/threonine kinase that is thought to be present in all eukarotic cells, implying that it has fundamental cellular functions.

Brand:  Invitrogen™ PR6299B

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15478463

  • 18250.00 SEK / 100µg
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The CSNK2A2 holoenzyme is a tetramer containing 2 alpha or alpha-prime subunits (or one of each) and 2 beta subunits. The function of beta subunits is unknown and the alpha subunit is the catalytic unit.

Specifications

Specifications

NP_001887
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
CSNK2A2 (CK2 α 2)
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
69.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
CK2 Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers