missing translation for 'onlineSavingsMsg'
Learn More

CstF-64 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26825-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16885175

  • 1355.00 SEK / 20µL
Estimated Shipment: 30-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
Specifications

Specifications

CstF-64
Polyclonal
Unconjugated
CSTF2
CF 1 64 kDa subunit, CstF 64, CSTF 64 kDa subunit, CSTF2, CstF-64
Rabbit
Antigen Affinity Chromatography
RUO
108062, 1478, 683927
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P33240, Q8BIQ5
Cstf2
CstF-64 Fusion Protein Ag25254
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers