missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CSTF1 (Human) Recombinant Protein
Human CSTF1 full-length ORF ( AAH01011, 1 a.a. - 431 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001477-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MYRTKVGLKDRQQLYKLIISQLLYDGYISIANGLINEIKPQSVCAPSEQLLHLIKLGMENDDTAVQYAIGRSDTVAPGTGIDLEFDADVQTMSPEASEYETCYVTSHKGPCRVATYSRDGQLIATGSADASIKILDTERMLAKSAMPIEVMMNETAQQNMENHPVIRTLYDHVDEVTCLAFHPTEQILASGSRDYTLKLFDYSKPSAKRAFKYIQEAEMLRSISFHPSGDFILVGTQHPTLRLYDINTFQCFVSCNPQDQHTDAICSVNYNSSANMYVTGSKDGCIKLWDGVSNRCITTFEKAHDGAEVCSAIFSKNSKYILSSGKDSVAKLWEISTGRTLVRYTGAGLSGRQVHRTQAVFNHTEDYVLLPDERTISLCCWDSRTAERRNLLSLGHNNIVRCIVHSPTNPGFMTCSDDFRARFWYRRSTTD
Specifications
AAH01011 | |
cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSTF1 | |
GST |
1477 | |
Wheat germ expression system | |
10 μg | |
CstF-50, CstFp50 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |