missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSTF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10064-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.Specifications
CSTF1 | |
Polyclonal | |
Unconjugated | |
CSTF1 | |
CF 1 50 kDa subunit, Cleavage stimulation factor, CstF 50, CSTF 50 kDa subunit, CSTF1, CstFp50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1477, 311670, 67337 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q05048, Q5BJQ6, Q99LC2 | |
Cstf1 | |
CSTF1 Fusion Protein Ag0109 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |