missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CSTF3 (Human) Recombinant Protein
Human CSTF3 full-length ORF ( AAH09792, 1 a.a. - 103 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001479-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSGDGATEQAAEYVPEKVKKAEKKLEENPYDLDAWSILIREAQNQPIDKARKTYERLVAQFPSSGRFWKLYIEAEVTILFYFFLYQYCSIHCSDRKQVRNIAN
Specifications
AAH09792 | |
cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSTF3 | |
GST |
1479 | |
Wheat germ expression system | |
10 μg | |
CSTF-77, MGC117398, MGC43001, MGC75122 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |