missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT83 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568001
Additional Details : Weight : 0.01000kg
Description
CT83 is a protein coding gene.Specifications
CT83 | |
Polyclonal | |
Unconjugated | |
CT83 | |
Cancer/testis antigen 83; CT83; CXorf61; kita-kyushu lung cancer antigen 1; KKLC1; KK-LC-1 | |
Rabbit | |
affinity chromatography | |
RUO | |
203413 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5H943 | |
CT83 | |
Synthesized peptide derived from human CT83. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |