Läs mer
Tocris Bioscience™ CTA 056
Beskrivning
CTA 056 is an interleukin-2-inducible T-cell kinase (ITK) inhibitor (IC50 values are 0.1, 0.4 and 5 μM for ITK, BTK and ETK, respectively). Inhibits growth of cancer cells expressing ITK, including MOLT-4 and Jurkat cells in vitro. Induces apoptosis in Jurkat cells. Inhibits MOLT-4 xenograft tumor growth in mice.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1,5-Dihydro-7-(phenylmethyl)-1-[3-(1-piperidinyl)propyl]-2-[4-(4-pyridinyl)phenyl]-6H-imidazo[4,5-g]quinoxalin-6-one |
| CAS | 1265822-30-7 |
| Kvantitet | 2 mg |
| Mål | ITK Inhibitors |
| Molekylformel | C35H34N6O |
| Renhet | 98% |