missing translation for 'onlineSavingsMsg'
Learn More

CTAGE5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55279-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826775

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to CTAGE5. It can recognize all the isoforms except isoform 5B.

CTAGE5 is a tumor-associated antigen.
Specifications

Specifications

CTAGE5
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15320
cTAGE family member 5, CTAGE family, member 5, CTAGE5, MEA11, MEA6, MGEA, MGEA11, MGEA6, Protein cTAGE 5
Rabbit
IgG
150 μL
-20°C
Primary
4253
Western Blot
Unconjugated
CTAGE5
Liquid
CTAGE5
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers